siRNA Details
| Gene Symbol: Tbc1d12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145952.3 | NP_666064.3 | 12, 13 | 2164 | ||
| GenBank mRNA | AK172979.1 | - | 12, 13 | 2158 | ||
| BC033574.1 | - | 3, 4 | 466 | |||
| BC138056.1 | - | 12, 13 | 2060 | 8861075 | ||
| BC138057.1 | - | 12, 13 | 2060 | 8861076 | ||
| BC145129.1 | - | 10, 11 | 1835 | 9053677 | ||
| BU152220.1 | - | 1, 2 | 241 | |||

Tbc1d12
Genomic Map