siRNA Details
| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145987.2 | NP_666099.2 | 3, 4 | 991 | ||
| GenBank mRNA | AK085940.1 | - | 991 | |||
| AK162322.1 | - | 3, 4 | 340 | |||
| BC025833.1 | - | 3, 4 | 392 | 5125918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem82| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145987.2 | NP_666099.2 | 3, 4 | 991 | ||
| GenBank mRNA | AK085940.1 | - | 991 | |||
| AK162322.1 | - | 3, 4 | 340 | |||
| BC025833.1 | - | 3, 4 | 392 | 5125918 | ||