siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145828.3 | NP_665827.2 | 4 | 1008 | ||
| GenBank mRNA | AJ291751.2 | - | 4 | 925 | ||
| AK083482.1 | - | 4 | 977 | |||
| AK086804.1 | - | 4 | 940 | |||
| AK150521.1 | - | 4 | 961 | |||
| AK153332.1 | - | 4 | 961 | |||
| AK158056.1 | - | 4 | 1008 | |||
| AK177270.1 | - | 1 | 26 | |||
| BC034082.1 | - | 3 | 527 | 4239646 | ||

Xylt2
Genomic Map