siRNA Details
| Gene Symbol: Vmn1r188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145850.1 | NP_665849.1 | 1 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r188| Gene Symbol: Vmn1r188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145850.1 | NP_665849.1 | 1 | 482 | ||