siRNA Details
| Gene Symbol: AW549877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145930.2 | NP_666042.2 | 4 | 495 | ||
| GenBank mRNA | AK045350.1 | - | 4 | 490 | ||
| AK154940.1 | - | 4 | 490 | |||
| BC016504.1 | - | 3 | 373 | 3709876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW549877| Gene Symbol: AW549877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145930.2 | NP_666042.2 | 4 | 495 | ||
| GenBank mRNA | AK045350.1 | - | 4 | 490 | ||
| AK154940.1 | - | 4 | 490 | |||
| BC016504.1 | - | 3 | 373 | 3709876 | ||