siRNA Details
| Gene Symbol: Zc3h7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145931.2 | NP_666043.2 | 5 | 603 | ||
| GenBank mRNA | AK028438.1 | - | 5 | 528 | ||
| AK040507.1 | - | 5 | 551 | |||
| AK049834.1 | - | 5 | 538 | |||
| AK052830.1 | - | 6 | 714 | |||
| AK150952.1 | - | 5 | 522 | |||
| AK161148.1 | - | 5 | 581 | |||
| AK166876.1 | - | 5 | 551 | |||
| AK167758.1 | - | 5 | 552 | |||
| BC027330.1 | - | 5 | 499 | 3674968 | ||
| BC029320.1 | - | 5 | 501 | |||

Zc3h7a
Genomic Map