siRNA Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145972.4 | NP_666084.1 | 3 | 373 | ||
| GenBank mRNA | AK137274.1 | - | 2 | 1066 | ||
| AK144612.1 | - | 3 | 354 | |||
| AK162291.1 | - | 2 | 215 | |||
| BB651637.1 | - | 3 | 371 | |||
| BC027231.1 | - | 3 | 376 | 3585175 | ||

BC027231
Genomic Map