siRNA Details
| Gene Symbol: 8430408G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166580.1 | NP_001160052.1 | 2 | 802 | ||
| NM_145980.2 | NP_666092.1 | 2 | 805 | |||
| GenBank mRNA | AB024924.1 | - | 2 | 731 | ||
| AK078806.1 | - | 2 | 807 | |||
| AK087284.1 | - | 2 | 804 | |||
| BC022212.1 | - | 2 | 778 | |||
| BC031533.1 | - | 2 | 790 | 2649431 | ||
| BC058515.1 | - | 2 | 773 | 6820680 | ||

8430408G22Rik
Genomic Map