siRNA Details
| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145987.2 | NP_666099.2 | 4 | 1006 | ||
| GenBank mRNA | AK085940.1 | - | 1006 | |||
| AK154809.1 | - | 3 | 1065 | |||
| AK162322.1 | - | 4 | 355 | |||
| BC025833.1 | - | 4 | 407 | 5125918 | ||

Tmem82
Genomic Map