siRNA Details
| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145838.1 | NP_665837.1 | 8 | 3019 | ||
| GenBank mRNA | AB059554.1 | - | 8 | 3019 | ||
| AK085105.1 | - | 9 | 3156 | |||
| AK156557.1 | - | 8 | 3559 | |||
| AK172400.1 | - | 8 | 3085 | |||

St8sia6
Genomic Map