siRNA Details
| Gene Symbol: Vmn1r69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145842.3 | NP_665841.1 | 1 | 1193 | ||
| GenBank mRNA | AF394954.1 | - | 1 | 1192 | ||
| Gene Symbol: Vmn1r68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172072.1 | NP_001165543.1 | 1 | 508 | ||

Vmn1r69
Genomic Map