siRNA Details
| Gene Symbol: Vmn1r192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145845.1 | NP_665844.1 | 1 | 780 | ||
| GenBank mRNA | BC138395.1 | - | 1 | 873 | 8861415 | |
| BC138400.1 | - | 1 | 872 | 8861420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r192| Gene Symbol: Vmn1r192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145845.1 | NP_665844.1 | 1 | 780 | ||
| GenBank mRNA | BC138395.1 | - | 1 | 873 | 8861415 | |
| BC138400.1 | - | 1 | 872 | 8861420 | ||