siRNA Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043344.1 | - | 22 | 2391 | ||
| AK085048.1 | - | 1548 | ||||
| AK145200.1 | - | 23 | 2447 | |||
| AK161473.1 | - | 23 | 2530 | |||
| BC024848.1 | - | 6 | 1043 | |||
| BC030328.1 | - | 23 | 2447 | 5376020 | ||

Cenpi
Genomic Map