siRNA Details
| Gene Symbol: Tspan14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145928.1 | NP_666040.1 | 9 | 2068 | ||
| GenBank mRNA | AK030593.1 | - | 9 | 2070 | ||
| AK156671.1 | - | 9 | 2081 | |||
| BC024611.1 | - | 9 | 2073 | 4188261 | ||
| BC025568.1 | - | 9 | 2082 | 5322531 | ||
| BC026574.1 | - | 9 | 2068 | 4954874 | ||

Tspan14
Genomic Map