siRNA Details
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145946.2 | NP_666058.2 | 11, 12 | 1056 | ||
GenBank mRNA | AK085572.1 | - | 9, 10 | 906 | ||
BC027836.1 | - | 11, 12 | 1052 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145946.2 | NP_666058.2 | 11, 12 | 1056 | ||
GenBank mRNA | AK085572.1 | - | 9, 10 | 906 | ||
BC027836.1 | - | 11, 12 | 1052 |