siRNA Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145958.2 | NP_666070.2 | 4 | 1446 | ||
| GenBank mRNA | AK028396.1 | - | 4 | 1392 | ||
| AK129374.1 | - | 4 | 1356 | |||
| BC022962.1 | - | 1 | 363 | 5368323 | ||
| BC023724.1 | - | 1 | 235 | |||

Kbtbd2
Genomic Map