siRNA Details
| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013276.2 | NP_037408.2 | 7 | 3681 | ||
| GenBank mRNA | AF163573.1 | - | 7 | 3681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHPK| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013276.2 | NP_037408.2 | 7 | 3681 | ||
| GenBank mRNA | AF163573.1 | - | 7 | 3681 | ||