siRNA Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 5 | 520 | ||
| GenBank mRNA | AK087586.1 | - | 5 | 518 | ||
| AK147246.1 | - | 5 | 476 | |||
| AK147269.1 | - | 5 | 476 | |||
| AK166346.1 | - | 5 | 489 | |||
| BC017516.1 | - | 5 | 513 | 4504441 | ||

Hip1
Genomic Map