siRNA Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271356.1 | NP_001258285.1 | 4 | 766 | ||
| NM_001271357.1 | NP_001258286.1 | 4 | 509 | |||
| NM_146018.2 | NP_666130.1 | 4 | 806 | |||
| GenBank mRNA | AK039106.1 | - | 4 | 593 | ||
| AK080933.1 | - | 4 | 591 | |||
| AK136071.1 | - | 4 | 497 | |||
| AK148216.1 | - | 4 | 560 | |||
| AK154040.1 | - | 4 | 788 | |||
| AK161460.1 | - | 4 | 538 | |||
| AK172245.1 | - | 4 | 565 | |||
| BC025820.1 | - | 4 | 540 | 5099493 | ||
| CJ052419.1 | - | 4 | 544 | |||

Flcn
Genomic Map