siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146067.3 | NP_666179.2 | 3 | 780 | ||
| GenBank mRNA | AK032288.1 | - | 3 | 743 | ||
| AK041329.1 | - | 3 | 753 | |||
| AK049721.1 | - | 3 | 781 | |||
| AK080876.1 | - | 3 | 738 | |||
| AK133977.1 | - | 3 | 774 | |||
| BC025476.1 | - | 3 | 701 | |||
| BC034347.1 | - | 3 | 422 | 2654874 | ||

Cpped1
Genomic Map