siRNA Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146068.4 | NP_666180.2 | 2 | 245 | ||
| GenBank mRNA | AK044836.1 | - | 2 | 228 | ||
| AK052832.1 | - | 2 | 210 | |||
| AK075871.1 | - | 2 | 210 | |||
| AK146053.1 | - | 2 | 245 | |||
| BC089026.1 | - | 2 | 182 | 30615298 | ||

Spidr
Genomic Map