siRNA Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146073.3 | NP_666185.3 | 2, 3 | 1253 | ||
| GenBank mRNA | AF542387.1 | - | 3, 4 | 922 | ||
| AK041496.1 | - | 2, 3 | 936 | |||
| AK041648.1 | - | 3, 4 | 1249 | |||
| AK046719.1 | - | 2, 3 | 1241 | |||
| AK080845.1 | - | 2, 3 | 1257 | |||
| BC059814.1 | - | 2, 3 | 1296 | 6813525 | ||
| BC086758.1 | - | 3, 4 | 309 | |||

Zdhhc14
Genomic Map