siRNA Details
| Gene Symbol: Fam13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146084.1 | NP_666196.1 | 3 | 595 | ||
| GenBank mRNA | AK029766.1 | - | 3 | 587 | ||
| AK054416.1 | - | 1 | 1001 | |||
| AK084711.1 | - | 3 | 604 | |||
| BC031465.1 | - | 3 | 595 | 3981146 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106158, XM_008772030, XM_008772031, XM_008772032, XM_008772033, and others.

Fam13b
Genomic Map