siRNA Details
| Gene Symbol: Tmem185b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146103.2 | NP_666215.1 | 1 | 778 | ||
| GenBank mRNA | AK010846.1 | - | 1 | 506 | ||
| AK031130.1 | - | 1 | 774 | |||
| BC020408.1 | - | 1 | 766 | |||
| BC024712.1 | - | 1 | 765 | 2646072 | ||

Tmem185b
Genomic Map