siRNA Details
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146105.3 | NP_666217.2 | 9 | 1369 | ||
| GenBank mRNA | AK036332.1 | - | 9 | 1401 | ||
| AK146681.1 | - | 9 | 1459 | |||
| BC034107.1 | - | 2 | 387 | 4503249 | ||
| BC039206.1 | - | 7 | 732 | |||
| BC054802.1 | - | 9 | 1369 | 6837803 | ||

Cnst
Genomic Map