siRNA Details
| Gene Symbol: BC025446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC025446.1 | - | 4 | 534 | 5053382 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC025446| Gene Symbol: BC025446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC025446.1 | - | 4 | 534 | 5053382 | |