siRNA Details
| Gene Symbol: Nrros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146069.4 | NP_666181.2 | 4 | 1775 | ||
| GenBank mRNA | AK028367.1 | - | 2 | 727 | ||
| AK042502.1 | - | 2 | 577 | |||
| AK084858.1 | - | 4 | 1778 | |||
| AK088770.1 | - | 2 | 496 | |||
| AK128897.1 | - | 3 | 615 | |||
| AK167934.1 | - | 3 | 645 | |||
| BC027411.1 | - | 3 | 625 | 4192329 | ||

Nrros
Genomic Map