siRNA Details
| Gene Symbol: Muc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145874.1 | NP_001139346.1 | 2 | 371 | ||
| NM_146071.2 | NP_666183.2 | 2 | 371 | |||
| GenBank mRNA | AB098732.1 | - | 2 | 386 | ||
| AK085640.1 | - | 2 | 338 | |||
| AK142548.1 | - | 2 | 372 | |||
| AK165477.1 | - | 2 | 363 | |||
| BB661244.1 | - | 2 | 372 | |||
| BC026367.1 | - | 2 | 372 | 4037422 | ||

Muc20
Genomic Map