siRNA Details
| Gene Symbol: Ppp4r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114131.1 | NP_001107603.1 | 11 | 1398 | ||
| NM_146081.2 | NP_666193.2 | 11 | 1449 | |||
| GenBank mRNA | AK133969.1 | - | 11 | 1320 | ||
| AK166418.1 | - | 11 | 1356 | |||
| BC026489.1 | - | 3 | 422 | 4504305 | ||
| BC029761.1 | - | 11 | 1384 | 3989055 | ||

Ppp4r1
Genomic Map