siRNA Details
| Gene Symbol: BC027072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146082.3 | NP_666194.2 | 1 | 2888 | ||
| GenBank mRNA | AK044416.1 | - | 1 | 497 | ||
| BC027072.1 | - | 1 | 469 | |||
| BC046516.1 | - | 1 | 2888 | 5364819 | ||
| BC060373.1 | - | 1 | 2897 | 6494066 | ||

BC027072
Genomic Map