siRNA Details
| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146108.1 | NP_666220.1 | 8 | 610 | ||
| GenBank mRNA | AK076038.1 | - | 8 | 609 | ||
| AK082965.1 | - | 8 | 609 | |||
| BC026437.1 | - | 8 | 610 | 4238681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hibch| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146108.1 | NP_666220.1 | 8 | 610 | ||
| GenBank mRNA | AK076038.1 | - | 8 | 609 | ||
| AK082965.1 | - | 8 | 609 | |||
| BC026437.1 | - | 8 | 610 | 4238681 | ||