siRNA Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 15 | 1474 | ||
| GenBank mRNA | AK147246.1 | - | 15 | 1430 | ||
| AK147269.1 | - | 15 | 1430 | |||
| AK166346.1 | - | 15 | 1443 | |||
| AK220182.1 | - | 2 | 175 | |||
| BC017516.1 | - | 15 | 1467 | 4504441 | ||

Hip1
Genomic Map