siRNA Details
Gene Symbol: Gm21975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146023.4 | NP_666135.1 | 4 | 1298 |
Gene Symbol: Evi2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077496.1 | NP_001070964.1 | 2 | 863 | ||
GenBank mRNA | AK039968.1 | - | 2 | 840 | ||
AK170476.1 | - | 2 | 825 | |||
AK171686.1 | - | 2 | 780 | |||
BC017548.1 | - | 2 | 812 | 4922351 |
