siRNA Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146027.2 | NP_666139.1 | 8 | 1482 | ||
| GenBank mRNA | AK075869.1 | - | 8 | 1446 | ||
| BC010784.1 | - | 7 | 1526 | |||
| BC021346.1 | - | 8 | 1451 | 4163541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scrn2| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146027.2 | NP_666139.1 | 8 | 1482 | ||
| GenBank mRNA | AK075869.1 | - | 8 | 1446 | ||
| BC010784.1 | - | 7 | 1526 | |||
| BC021346.1 | - | 8 | 1451 | 4163541 | ||