siRNA Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146087.2 | NP_666199.1 | 9, 10 | 1147 | ||
| GenBank mRNA | AK076070.1 | - | 9, 10 | 1414 | ||
| AK076264.1 | - | 9, 10 | 1406 | |||
| AK147165.1 | - | 9, 10 | 1392 | |||
| AK152387.1 | - | 9, 10 | 1392 | |||
| AK159939.1 | - | 6, 7 | 616 | |||
| AK166318.1 | - | 9, 10 | 1407 | |||
| AK166429.1 | - | 9, 10 | 1389 | |||
| AK191355.1 | - | 1, 2 | 135 | |||
| AK191813.1 | - | 1, 2 | 135 | |||
| AK192016.1 | - | 1, 2 | 135 | |||
| AK201811.1 | - | 1, 2 | 135 | |||
| AK207201.1 | - | 1, 2 | 135 | |||
| AK212753.1 | - | 1, 2 | 135 | |||
| BC019740.1 | - | 9, 10 | 1147 | 3487913 | ||
| BC025439.1 | - | 9, 10 | 1094 | 5038433 | ||
| BC048081.1 | - | 9, 10 | 1180 | 6309504 | ||
| X90945.1 | - | 1, 2 | 86 | |||

Csnk1a1
Genomic Map