siRNA Details
| Gene Symbol: Zadh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146090.5 | NP_666202.2 | 2 | 3188 | ||
| GenBank mRNA | AK081988.1 | - | 2 | 3188 | ||
| BC024490.1 | - | 1 | 927 | |||
| BC025897.1 | - | 1 | 2431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zadh2| Gene Symbol: Zadh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146090.5 | NP_666202.2 | 2 | 3188 | ||
| GenBank mRNA | AK081988.1 | - | 2 | 3188 | ||
| BC024490.1 | - | 1 | 927 | |||
| BC025897.1 | - | 1 | 2431 | |||