siRNA Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243741.1 | NP_001230670.1 | 3 | 1261 | ||
| NM_001243742.1 | NP_001230671.1 | 3 | 797 | |||
| NM_146170.4 | NP_666282.3 | 3 | 738 | |||
| GenBank mRNA | AK047127.1 | - | 3 | 739 | ||
| AK077101.1 | - | 564 | ||||
| AK077503.1 | - | 3 | 1148 | |||
| AK170524.1 | - | 4 | 629 | |||
| AK171511.1 | - | 4 | 629 | |||
| BC025632.1 | - | 3 | 705 | 5321669 | ||
| BC026926.1 | - | 3 | 808 | 3586097 | ||
| BC031913.1 | - | 3 | 751 | 5376246 | ||

C87436
Genomic Map