siRNA Details
| Gene Symbol: Zfp553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146201.1 | NP_666313.1 | 2 | 129 | ||
| GenBank mRNA | AK049208.1 | - | 2 | 511 | ||
| AK140924.1 | - | 2 | 332 | |||
| AK144911.1 | - | 2 | 342 | |||
| AK146581.1 | - | 2 | 349 | |||
| AK148378.1 | - | 2 | 280 | |||
| BC026401.1 | - | 2 | 129 | 4209324 | ||

Zfp553
Genomic Map