siRNA Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146207.2 | NP_666319.2 | 6 | 600 | ||
| GenBank mRNA | AK144802.1 | - | 6 | 582 | ||
| AK150237.1 | - | 6 | 579 | |||
| AK166251.1 | - | 6 | 582 | |||
| AK168215.1 | - | 6 | 582 | |||
| AK170722.1 | - | 6 | 582 | |||
| AK189560.1 | - | 2 | 86 | |||
| BC010211.1 | - | 6 | 578 | 3969515 | ||
| BC024113.1 | - | 2 | 72 | 5097944 | ||

Cul4a
Genomic Map