siRNA Details
| Gene Symbol: Klhl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146219.1 | NP_666331.1 | 3 | 459 | ||
| GenBank mRNA | AK131135.1 | - | 3 | 384 | ||
| AK157027.1 | - | 3 | 493 | |||
| AK164240.1 | - | 3 | 499 | |||
| BC025816.1 | - | 3 | 459 | 5098064 | ||

Klhl36
Genomic Map