siRNA Details
| Gene Symbol: Zfp426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110309.1 | NP_001103779.1 | 5, 6 | 436 | ||
| NM_146221.4 | NP_666333.1 | 3, 4 | 574 | |||
| GenBank mRNA | AK036912.1 | - | 5, 6 | 328 | ||
| AK085144.1 | - | 3, 4 | 575 | |||
| AK220563.1 | - | 3, 4 | 1276 | |||
| BC023931.1 | - | 5, 6 | 441 | 5324400 | ||
| BC024805.1 | - | 5, 6 | 432 | 5359610 | ||

Zfp426
Genomic Map