siRNA Details
| Gene Symbol: Fam129b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146119.2 | NP_666231.1 | 4 | 593 | ||
| GenBank mRNA | AK033735.1 | - | 4 | 561 | ||
| AK046583.1 | - | 4 | 490 | |||
| AK051396.1 | - | 4 | 532 | |||
| AK146953.1 | - | 4 | 578 | |||
| AK149888.1 | - | 4 | 503 | |||
| AK150642.1 | - | 4 | 531 | |||
| AK152207.1 | - | 4 | 552 | |||
| AK153489.1 | - | 4 | 537 | |||
| AK170512.1 | - | 4 | 533 | |||
| BC027843.1 | - | 4 | 513 | 5343013 | ||

Fam129b
Genomic Map