siRNA Details
| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146162.2 | NP_666274.1 | 2 | 836 | ||
| GenBank mRNA | AK078473.1 | - | 2 | 845 | ||
| AK135223.1 | - | 867 | ||||
| AK170712.1 | - | 2 | 845 | |||
| BC025600.1 | - | 2 | 841 | 5250899 | ||

Tmem119
Genomic Map