siRNA Details
| Gene Symbol: Tubgcp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146190.2 | NP_666302.2 | 11 | 1285 | ||
| GenBank mRNA | AK032031.1 | - | 11 | 1245 | ||
| AK034715.1 | - | 2 | 3364 | |||
| AK044444.1 | - | 11 | 1171 | |||
| AK044887.1 | - | 11 | 1269 | |||
| AK051326.1 | - | 11 | 1280 | |||
| AK086158.1 | - | 11 | 1246 | |||
| AK134963.1 | - | 3 | 403 | |||
| AK151454.1 | - | 11 | 1243 | |||
| AK152033.1 | - | 11 | 1243 | |||
| AK153331.1 | - | 11 | 1243 | |||
| AK153357.1 | - | 11 | 1243 | |||
| AK160939.1 | - | 11 | 1199 | |||
| AK161681.1 | - | 11 | 1243 | |||
| AK169443.1 | - | 2 | 629 | |||
| AK173291.1 | - | 8 | 662 | |||
| BC027075.1 | - | 3 | 348 | 5355335 | ||

Tubgcp5
Genomic Map