siRNA Details
| Gene Symbol: Zfp553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146201.1 | NP_666313.1 | 2 | 341 | ||
| GenBank mRNA | AK049208.1 | - | 2 | 723 | ||
| AK140924.1 | - | 2 | 544 | |||
| AK144911.1 | - | 2 | 554 | |||
| AK146581.1 | - | 2 | 561 | |||
| AK148378.1 | - | 2 | 492 | |||
| AK185381.1 | - | 1 | 71 | |||
| BC012403.1 | - | 1 | 202 | |||
| BC026401.1 | - | 2 | 341 | 4209324 | ||

Zfp553
Genomic Map