siRNA Details
| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146222.2 | NP_666334.2 | 4 | 928 | ||
| GenBank mRNA | AK153989.1 | - | 3 | 281 | ||
| BC024479.1 | - | 3 | 353 | 4977181 | ||
| BC066002.1 | - | 4 | 928 | 5701381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd2| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146222.2 | NP_666334.2 | 4 | 928 | ||
| GenBank mRNA | AK153989.1 | - | 3 | 281 | ||
| BC024479.1 | - | 3 | 353 | 4977181 | ||
| BC066002.1 | - | 4 | 928 | 5701381 | ||