siRNA Details
| Gene Symbol: Zfp280d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146224.4 | NP_666336.3 | 5 | 430 | ||
| GenBank mRNA | AK045173.1 | - | 5 | 326 | ||
| AK146597.1 | - | 5 | 326 | |||
| AK169622.1 | - | 5 | 423 | |||
| AK172652.1 | - | 5 | 422 | |||
| BB616601.1 | - | 5 | 430 | |||
| BC042445.1 | - | 4 | 290 | |||
| BC079878.1 | - | 5 | 342 | 6855096 | ||

Zfp280d
Genomic Map