siRNA Details
Gene Symbol: Slc22a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146232.1 | NP_666344.1 | 2 | 652 | ||
GenBank mRNA | AJ132857.1 | - | 563 | |||
BC014805.1 | - | 2 | 652 | 4242162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146232.1 | NP_666344.1 | 2 | 652 | ||
GenBank mRNA | AJ132857.1 | - | 563 | |||
BC014805.1 | - | 2 | 652 | 4242162 |