siRNA Details
| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301414.1 | NP_001288343.1 | 5 | 991 | ||
| NM_146144.4 | NP_666256.2 | 5 | 972 | |||
| GenBank mRNA | AK080882.1 | - | 5 | 973 | ||
| BC018179.1 | - | 5 | 852 | 3585191 | ||
| BC020007.1 | - | 5 | 824 | 3980469 | ||
| BC023812.1 | - | 2 | 146 | |||

Usp1
Genomic Map