siRNA Details
| Gene Symbol: C8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290645.1 | NP_001277574.1 | 10 | 3328 | ||
| NM_146148.2 | NP_666260.1 | 11 | 3425 | |||
| GenBank mRNA | AB077295.1 | - | 12 | 1796 | ||
| AB077296.1 | - | 11 | 3388 | |||
| AB077297.1 | - | 10 | 3289 | |||
| AB077298.1 | - | 11 | 3383 | |||
| AK090245.1 | - | 11 | 3427 | |||
| BC027748.1 | - | 11 | 3421 | 5052412 | ||

C8a
Genomic Map